Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18, APC, clone: 212701, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523529

Catalog No. PIMA523529

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD18, integrin beta2 subunit, forms heterodimers with four types of CD11 molecule to constitute leukocyte (beta2) integrins: alphaLbeta2 (CD11a/CD18, LFA-1), alphaMbeta2 (CD11b/CD18, Mac-1, CR3), alphaXbeta2 (CD11c/CD18) and alphaDbeta2 (CD11d/CD18). In most cases, the response mediated by the integrin is a composite of the functions of its individual subunits. These integrins are essential for proper leukocyte migration, mediating intercellular contacts. Absence of CD18 leads to leukocyte adhesion deficiency-1, severe reduction of CD18 expression leads to the development of a psoriasiform skin disease. CD18 is also a target of Mannheimia (Pasteurella) haemolytica leukotoxin and is sufficient to mediate leukotoxin-mediated cytolysis.

In direct ELISAs, no cross-reactivity with recombinant human Integrin beta 1 or recombinant mouse Integrin alpha 5 is observed.
Specifications

Specifications

CD18
Monoclonal
APC
ITGB2
Liquid
ITGB2
Mouse myeloma cell line NS0-derived recombinant human Integrin beta 2/CD18 Gln23-Asn700
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
212701
Buffered saline solution with BSA and 0.09% sodium azide
P05107
Cd18, LAD, LCAMB, Lfa1, MF17, Integrin beta-2 subunit, ITGB2, cluster designation 18
Murine
IgG1
100 Tests
Store at 4°C, store in the dark, do not freeze
Primary
3689
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit