Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD104 Rat anti-Mouse, Clone: 308601, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524024

Catalog No. PIMA524024


Add to cart

Description

Description

This antibody does not cross-react with recombit mouse Integrin beta 1, beta 2, beta 6, beta 7, or recombit human Integrin beta 3. Reconstitute at 0.5 mg/mL in sterile PBS.

Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications

Specifications

CD104
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
A2A863
CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, ITGB4
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
192897
Flow Cytometry, Immunohistochemistry, Western Blot
308601
Unconjugated
Itgb4
Lyophilized
ITGB4
CHO-derived recombit mouse Integrin beta 4/CD104 Asn29-Ser711
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit