Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB6 Mouse anti-Human, APC, Clone: 437211, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523675

Catalog No. PIMA523675


Add to cart

Description

Description

Integrin alpha-V/beta-6 is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalisation of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.
Specifications

Specifications

ITGB6
Monoclonal
APC
ITGB6
Liquid
ITGB6
CHO-derived recombit human Integrin alpha V beta 6 Phe31-Val992 (Integrin alpha V) and Gly22-Asn707 (Integrin beta 6)
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
437211
Buffered saline solution with BSA and 0.09% sodium azide
P18564
integrin beta 6
Mouse
IgG2B
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3694
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit