Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Integrin α M/CD11b, Clone: 238446, R&D Systems™

Mouse Monoclonal Antibody

Specifications

Antigen CD11b
Immunogen Mouse myeloma cell line NS0-derived recombinant human Integrin alpha M/CD11b, Phe17-Asn1105, Accession # NP_001139280
Clone 238446
Applications Flow Cytometry
Conjugate Alexa Fluor 405
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB16991V02
View Documents
R&D Systems
FAB16991V025
25 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FAB16991V10
View Documents
R&D Systems
FAB16991V100
100 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

CD11b
238446
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Mouse
IgG2b
3684
RUO
Mouse myeloma cell line NS0-derived recombinant human Integrin alpha M/CD11b, Phe17-Asn1105, Accession # NP_001139280
Flow Cytometry
Supernatant
NP_001139280
Monoclonal
Primary
Detects human Integrin alpha M/CD11b in direct ELISAs.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit