Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αVΒ6 Mouse anti-Human, Clone: E7P6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2074Z
Description
This Anti-Integrin αVβ6 Antibody, clone E7P6, azide free is validated for use in flow cytometry, immunocytochemistry, immunohistochemistry for the detection of Integrin αVβ6.
Specifications
Integrin αVβ6 | |
Monoclonal | |
Unconjugated | |
NM_002210.2 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
E7P6 | |
Sterile-filtered liquid at 1mg/mL in PBS, containing no preservatives. | |
ITGAV, DKFZp686A08142, MSK8, CD51, VNRA | |
Recombinant secreted human alphavbeta6. | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction