Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intelectin 2 Rat anti-Mouse, Clone: 747421, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524358

Catalog No. PIMA524358


Add to cart

Description

Description

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Specifications

Specifications

Intelectin 2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q80ZA0
HL-2, Intelectin-b, Itln1b, ITLN2, Itlnb, HL-2, intelectin 2, UNQ2789/PRO7179
Rat
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
493583
Immunohistochemistry (Frozen), Western Blot
747421
Unconjugated
Itlnb
Lyophilized
Itlnb
E. coli-derived recombit mouse Intelectin-2 Ala20-Ser298
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit