Learn More
Intercalated DNA Mouse anti-Many, Clone: ET749.4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110599
Catalog No. 01-671-552
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10599 detects Intercalated DNA from many eukaryotic DNA samples. MA1-10599 has been successfully used in ELISA applications. The MA1-10599 immunogen is Ethidium bromide intercalated calf thymus DNA.
Specifications
Specifications
Intercalated DNA
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Many
ELISA
ET749.4
Unconjugated
Liquid
Ethidium bromide intercalated calf thymus DNA.
Protein G
RUO
Antibody
Monoclonal