Learn More
Interferon Beta Rabbit anti-Human, Bovine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061095
Catalog No. 06-109-5MI
Add to cart
Description
Description
Detect Interferon Beta using this Anti-Interferon Beta Antibody validated for use in western blot, NEUT, immunohistochemistry (paraffin).
Specifications
Specifications
Interferon-β
Unconjugated
Serum
Rabbit
Unpurified
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Bovine, Human
Immunohistochemistry (Paraffin), Neutralization, Western Blot
Rabbit polyclonal containing serum with 0.05% sodium azide.
IFNB1; IFB; IFF; IFNB
KLH-conjugated linear peptide corresponding to the internal domain of human Interferon beta.
200μL
Primary
NP_002167