Learn More
Interferon alpha-6 Mouse anti-Human, Clone: 3C9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519186
Catalog No. PIMA519186
Add to cart
Description
Description
Peptide Sequence: WDERLLDKLY TELYQQLNDL EACVMQEVWV GGTPLMNEDS ILAVRKYFQR ITLYLTEKKY SPCAWEVVRA EIMRSFSSSR NLQERLRRKE
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
Specifications
Interferon alpha-6
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P05013
RP11-354P17.7, IFN-alphaK, IFN-alpha-6, interferon alpha-54, interferon alpha-6, interferon alpha-K, leIF K
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3443
ELISA, Immunoprecipitation, Western Blot
3C9
Unconjugated
IFNA6
Liquid
IFNA6
IFNA6 (NP_066282, 100 a.a. ∽ 189 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human