Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon alpha-8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521679

Catalog No. PIPA521679


Add to cart

Description

Description

PA5-21679 targets Interferon alpha-8 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-21679 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 189 of interferon alpha 8 (Uniprot IDNo.P32881). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Interferon alpha-8
Polyclonal
Unconjugated
IFNA8
Liquid
IFNA8
Recombit fragment corresponding to a region within amino acids 1 and 189 of interferon alpha 8 (Uniprot IDNo.P32881)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P32881
RP11-354P17.14-001, IFN-alphaB, IFN-alpha-8, interferon alpha type 201, interferon alpha-8, interferon alpha-B, interferon alpha-B′, interferon alpha-B2, leIF B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3445
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit