Learn More
IFN beta Sheep anti-Human, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA128044
Catalog No. PIPA128044
Add to cart
Description
Description
PA1-28044 detects IFN beta from human samples. PA1-28044 has been successfully used in control applications. The PA1-28044 immunogen is: Recombit full length IFN beta derived from E. Coli (Human).
Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
Specifications
Specifications
IFN beta
Polyclonal
whole serum diluted in PBS with no preservative
P01574
IFNB1
Recombit full length IFN beta derived from E. Coli (Human).
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3456
Control, Functional Assay, Neutralization
Unconjugated
IFNB1
Liquid
Sheep
IgG
RUO
Antibody
Polyclonal
Human