Learn More
Interleukin 13 (IL-13) Rabbit anti-Human, Rhesus Macaque, Chimpanzee, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061090
Catalog No. 06-109-0MI
Add to cart
Description
Description
Detect Interleukin 13 (IL-13) using this Anti-Interleukin 13 (IL-13) Antibody validated for use in western blot, immunohistochemistry.
Specifications
Specifications
Interleukin 13 (IL-13)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Chimpanzee, Human, Rhesus
Immunohistochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
IL13; ALRH; IL-13; P600; BHR1
KLH-conjugated linear peptide corresponding to the mature chain of Interleukin-13 (IL-13).
100μg
Primary
NP_002179