Learn More
Interleukin 17 Receptor Beta (IL-17RB) Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061071
Catalog No. 06-107-1MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-Interleukin 17 Receptor Beta (IL-17RB) Antibody is validated for use in western blot, FC for the detection of Interleukin 17 Receptor Beta (IL-17RB).
Specifications
Specifications
Interleukin 17 Receptor Beta (IL-17RB)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human, Murine
Flow Cytometry, Western Blot
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.
IL17RB1/27/201; CRL4; EVI27; IL-17RB; IL-17Rh1; IL17BR; IL17RH1; IL17Rh1
KLH-conjugated linear peptide corresponding to the extracellular domain of human IL-17 receptor B.
100μg
Primary
NP_062529