Learn More
Interleukin 21 (IL-21) Rabbit anti-Human, Mouse, Monkey, Chimpanzee, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061074
Catalog No. 06-107-4MI
Add to cart
Description
Description
Anti-Interleukin 21 (IL-21) Antibody is an antibody against Interleukin 21 (IL-21) for use in flow cytometry, immunohistochemistry (paraffin) and western blotting.
Specifications
Specifications
Interleukin 21 (IL-21)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Chimpanzee, Human, Monkey, Murine
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Purified rabbit polyclonal in PBS with 0.06% sodium azide.
IL21; IL-21
KLH-conjugated linear peptide corresponding to the C-terminus of human IL-21.
100μg
Primary
NP_068575