Learn More
Interleukin 23 alpha (IL-23A) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061079
Catalog No. 06-107-9MI
Add to cart
Description
Description
Detect Interleukin 23 alpha (IL-23A) using this Anti-Interleukin 23 alpha (IL-23A) Antibody validated for use in western blot, flow cytometry.
Specifications
Specifications
Interleukin 23 alpha (IL-23A)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human, Murine
Flow Cytometry, Western Blot
Purified rabbit polyclonal in PBS with 0.09% sodium azide.
IL23A; IL-23; IL-23A; IL-23p19; IL23P19; P19; SGRF
KLH-conjugated linear peptide corresponding to the mature chain of human IL-23 subunit alpha.
100μg
Primary
NP_057668