Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intestinal Alkaline Phosphatase Rabbit anti-Human, Biotin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126872

Catalog No. PIPA126872


Add to cart

Description

Description

PA1-26872 detects human intestinal Alkaline Phosphatase from human samples. PA1-26872 has been successfully used in ELISA, immunomicroscopy and Western blot procedures. The PA1-26872 immunogen is Alkaline Phosphatase from human intestine.

Specifications

Specifications

Intestinal Alkaline Phosphatase
Polyclonal
Biotin
ALPI
Liquid
Rabbit
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
248
ELISA, Immunomicroscopy, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2
P09923
ALPI
Alkaline Phosphatase from human intestine.
IgG fraction
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit