Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL10 Mouse anti-Human, PE, Clone: 33036, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523625

Catalog No. PIMA523625

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

The chemokine IP-10 (or CXCL10) is a chemokine made by monocytes, endothelial cells and fibroblasts in response to treatment with IFNgamma. IP-10 functions as a chemoattractant to cells expressing the G protein-coupled receptor, CXCR3, which is found mainly on activated T cells and NK cells. IP-10 plays an important role in Th1 type inflammatory diseases and autoimmune diseases such as, Hashimoto ft.s thyroiditis, Graves ft. disease and Type 1 diabetes mellitus.
Specifications

Specifications

CXCL10
Monoclonal
PE
CXCL10
Liquid
CXCL10
E. coli-derived recombit human CXCL10/IP-10/CRG-2 Val22-Pro98
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
33036
Buffered saline solution with BSA and 0.09% sodium azide
P02778
C-X-C motif chemokine 10, Interferon Gamma Inducible Protein 10, C7, CRG-2, INP10, Ifi10, Scyb10, gIP-10, mob-1
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3627
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit