Learn More
IPS-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061096
Catalog No. 06-109-6MI
Add to cart
Description
Description
Anti-IPS-1 Antibody is an antibody against IPS-1 for use in western blot, immunohistochemistry.
Specifications
Specifications
IPS-1
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human
Immunohistochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
MAVS; CARDIF; Cardif; IPS-1; IPS1; VISA
KLH-conjugated linear peptide corresponding to human IPS-1 at and around the cytoplasmic domain.
100μg
Primary
NP_065797