Learn More
IQGAP1 Mouse, Unlabeled, Clone: 24, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610612
Description
GTPases (Ras, Rho, cdc42Hs, and rac) modulate various cellular functions such as cytoskeletal architecture, growth, motility, and gene expression. The activity of the GTP-binding proteins is regulated by factors that accelerate GTP-binding (GAPs) and proteins that enhance the rate of GTP hydrolysis (GTPases). IQGAP1 is Ras-GAP related protein that contains a WW domain that binds poly-proline regions, a calponin-homology domain (CHD), an IQ domain of unconventional myosins, and a C-terminal GAP-related domain (GRD). IQGAP1 co-immunoprecipitates with cdc42 and calmodulin, and colocalizes with cdc42Hs at the leading edges of the plasma membrane and at the cell-cell junctions. IQGAP1 may interact with both cadherins and catenins leading to dissociation of these proteins from sites of cell adhesion. Thus, IQGAP1 may be a Rac1 and Cdc42 effector that negatively regulates cadherin-mediated cell junctions.
Specifications
IQGAP1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
24 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.