Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IQGAP2 Mouse anti-Human, Mouse, Clone: BB9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05505
Description
Anti-IQGAP2 Antibody, clone BB9 detects level of IQGAP2 and has been published and validated for use in immunoprecipitation and western blotting.
Specifications
IQGAP2 | |
Monoclonal | |
Unconjugated | |
NM_006633 | |
Mouse | |
Ascites | |
Primary | |
−20°C for 24 months. | |
IgG |
Immunoprecipitation, Western Blot | |
BB9 | |
0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide containing 30% glycerol | |
IQGAP2 | |
GST-fusion protein corresponding to residues 519-939 of human IQGAP2, purified from E. coli | |
200 μL | |
Human, Mouse | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction