Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141596

Catalog No. 01-675-213


Add to cart

Description

Description

Suggested positive control: Jurkat whole cell lysate.

IRAK2 (interleukin-1 receptor-associated kinase-like 2) binds to the IL-1 type I receptor following IL-1 engagment and triggers intracellular signaling cascades leading to transcriptional up-regulation and mRNA stabilization. The pro-inflammatory cytokine IL-1 induces cellular response through two subunits of its receptor, IL-1 receptor I (IL-1RI) and IL-1 receptor accessory protein (IL-1RAcP). IL-1 receptor-associated kinase (IRAK) mediates activation of NF-kappa-B , which is a pivotal transcription factor mediating inflammatory and immune response. A novel member in the IRAK/Pelle family was recently identified and designated IRAK2. Both IRAK and IRAK2 recruit to the subunits of the IL-1R complex after IL-1 binding and lead to NF-kappa-B activation. IRAKs also associate with Toll-like receptor (TLR) and the domit negative mutants of IRAKs inhibit LPS-induced NF-kappa-B activation. Members in IRAK/Pelle family play a central role in IL-1R and TLR mediated inflammatory responses to cytokine IL-1 and LPS. IRAK2 is expressed in a variety of human tissues.
Specifications

Specifications

IRAK2
Polyclonal
Unconjugated
IRAK2
Liquid
IRAK2
Synthetic peptide corresponding to a portion of human IRAK-2.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
O43187, Q8CFA1
IRAK-2, MGC150550
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
108960, 3656
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit