Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4, Mouse, Clone: A8A8, Abnova™

Mouse monoclonal antibody raised against synthetic phosphopeptide of IRAK4.

Manufacturer:  Abnova Corporation MAB2538

Catalog No. 89-024-823


Add to cart

Description

Description

This gene encodes a kinase that activates NF-kappaB in both the Toll-like receptor (TLR) and T-cell receptor (TCR) signaling pathways. The protein is essential for most innate immune responses. Mutations in this gene result in IRAK4 deficiency and recurrent invasive pneumococcal disease. Multiple transcript variants encoding different isoforms have been found for this gene. [supplied by RefSeq

Sequence: VMTSR
Specifications

Specifications

IRAK4
A8A8
Mouse monoclonal antibody raised against synthetic phosphopeptide of IRAK4.
In TBS, pH 7.2 (BSA, 10% Proclin300)
Liquid
IRAK4
Synthetic phosphopeptide corresponding to residues surrounding T345 of human IRAK4.
100 ug
Store at -20°C to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody recognizes IRAK4 (phospho T345) with a phosphorylated site at Threonine 345. It does not cross-react with non-phosphospecific peptide.
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
IRAK4
IPD1|NY-REN-64|REN64
Murine
IgG1, kappa
RUO
Primary
51135
Bovine, Canine, Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit