Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF1 Mouse anti-Human, Unlabeled, Clone: 20, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612047

 View more versions of this product

Catalog No. BDB612047


Add to cart

Description

Description

Cytokine function is critical for both innate and adaptive immune responses. Interferon (IFN) signal transduction pathways involve a variety of transcription factors that bind specific DNA motifs called IFN-stimulated response elements (ISRE) and IFN γ-activated sequences (GAS). The IFN regulatory factor (IRF) family is a group of transcription factors that bind to these DNA motifs and regulate the activity of IFN-responsive genes. The IRF family is comprised of seven different proteins (IRF1 to IRF7) that have been identified due to their interaction with ISRE and GAS sequences. IRF1 functions in the activation of IFN-responsive genes during induction of histocompatibility antigens, and inhibition of cell proliferation. In addition, IRF1 may function as a tumor suppressor, since deletions in the IRF1 locus is associated with myelodysplasia and leukemias. IRF2 is thought to be a transcriptional repressor, while others like IRF3 and IRF4 may function as activators or repressors depending on the specific promoter. In lymphocytes, IRF4 is required for normal development and function. Thus, IRFs regulate the activity of IFN-responsive genes required for normal immune function.

Host Species: Mouse
Clone: 20
Isotype: IgG1
Species Reactivity : Human
Immunogen: Human IRF1 aa. 159-279

Western Blotting

Specifications

Specifications

IRF1
20
Unconjugated
Affinity Purified
Human IRF1 aa. 159-279
150μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.