Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF2, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131652

Catalog No. PIPA131652

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

IRF2 encodes interferon regulatory factor 2, a member of the interferon regulatory transcription factor (IRF) family. IRF2 competitively inhibits the IRF1-mediated transcriptional activation of interferons alpha and beta, and presumably other genes that employ IRF1 for transcription activation. However, IRF2 also functions as a transcriptional activator of histone H4.

PA1-31652 detects IRF2 from human samples.PA1-31652 has been successfully used in Western blot applications.The PA1-31652 immunogen is: Peptide with sequence KTSDITQARVKSC, from C Terminus of the protein sequence according to NP_002190
Specifications

Specifications

IRF2
Polyclonal
Unconjugated
IRF2
Purified
IRF2
Peptide with sequence KTSDITQARVKSC, from C Terminus of the protein sequence according to NP_002190
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P14316, P23906
DKFZp686F0244, IRF-2
Goat
IgG
50μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3660
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit