Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF4 Rat anti-Human, Clone: 3E4, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma 04142
Description
Anti-IRF4 Antibody, clone 3E4 is a highly specific rat monoclonal antibody, that targets IRF4 and has been tested in western blotting, immunohistochemistry and Flow Cytometry.Specifications
IRF4 | |
Monoclonal | |
Unconjugated | |
NP_038702 | |
Rat | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Western Blot | |
3E4 | |
Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Irf4, Spip | |
GST-tagged recombinant protein corresponding to the C-terminus of mouse IRF4. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only