Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IRF5, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP178770

Catalog No. NBP178770

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IRF5
1.0 mg/mL
Western Blot 1:2500-1:25000, Immunoprecipitation 2-10 mcg/mg lysate
NP_036187.1
IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
IRF5
The immunogen for this product maps to a region between residue 175 and 225 of mouse Interferon Regulatory Factor 5 using the numbering given in entry NP_036187.1 (GeneID 27056).
Immunogen affinity purified
RUO
Primary
3663
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only