Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IRF5, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP131275

Catalog No. NBP131275

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IRF5
0.55mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
IRF5
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human IRF5. The exact sequence is proprietary.
56 kDa
0.1 ml
Apoptosis
Primary
3663
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only