Learn More
IRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06526
Catalog No. 06-526-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-IRS1 Antibody, pre-CT is an antibody against IRS1 for use in immunoprecipitation and western blotting.
Specifications
Specifications
IRS1
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_005544
Immunoprecipitation, Western Blot
Immunoaffinity Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, and 5mg/ml BSA before the addition of glycerol to 30%
IRS1; IRS-1; HIRS-1
KLH-linked, precarboxy-terminal 16 residue peptide corresponding to amino acid residues 1220-1235 of rat liver IRS-1
Protein A purfied
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine, Rat