Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Rabbit anti-Human, Mouse, Rat, Clone: AW58, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04784
Description
Anti-IRS1 Antibody, clone AW58 detects level of IRS1 and has been published and validated for use in immunoprecipitation and western blotting.Specifications
IRS1 | |
Monoclonal | |
Unconjugated | |
NM_005544 | |
Peptide corresponding to carboxy-terminal 14 amino acids (C-YASINFQKQPEDRQ) of rat liver IRS1 | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunoprecipitation, Western Blot | |
AW58 | |
Cultured supernantant in 0.05% sodium azide | |
Rabbit | |
100 μL | |
IRS1, IRS-1, HIRS-1 | |
−20°C for 24 months | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only