Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRX2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526500

Catalog No. PIPA526500

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

IRX2 is a member of the Iroquois homeobox gene family. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
Specifications

Specifications

IRX2
Polyclonal
PBS with 0.09% sodium azide
Q9BZI1, P81066
IRXA2, homeodomain protein IRXA2, iroquois homeobox protein 2, iroquois-class homeodomain protein IRX-2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
153572, 16372
Western Blot
Unconjugated
IRX2
Liquid
IRX2
KLH conjugated synthetic peptide between 180-208 amino acids from the central region of human IRX2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit