Learn More
iroquois homeobox 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human IRX3 protein.
Supplier: Abnova Corporation H00079191B01
Description
IRX3 is a member of the Iroquois homeobox gene family (see IRX1; MIM 606197) and plays a role in an early step of neural development (Bellefroid et al., 1998 [PubMed 9427753]). Members of this family appear to play multiple roles during pattern formation of vertebrate embryos (Lewis et al., 1999 [PubMed 10370142]).[supplied by OMIM
Sequence: MSFPQLGYQYIRPLYPSERPGAAGGSGGSAGARGGLGAGASELNASGSLSNVLSSVYGAPYAAAAAAAAAQGYGAFLPYAAELPIFPQLGAQYELKDSPGVQHPAAAAAFPHPHPAFYPYGQYQFGDPSRPKNATRESTSTLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRSRTDEEGNAYGSEREEEDEEEDEEDGKRELELEEEELGGEEEDTGGEGLADDDEDEEIDLENLDGAATEPELSLAGAARRDGDLGLGPISDSKNSDSEDSSEGLEDRPLPVLSLAPAPPPVAVASPSLPSPPVSLDPCAPAPAPASALQKPKIWSLAETATSPDNPRRSPPGAGGSPPGAAVAPSALQLSPAAAAAAAHRLVSAPLGKFPAWTNRPFPGPPPGPRPHPLSLLGSAPPHLLGLPGAAGHPAAAAAFARPAEPEGGTDRCSALEVEKKLLKTAFQPVPRRPQNHLDAALVLSALSSSSpecifications
iroquois homeobox 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human IRX3 protein. | |
IRX3 | |
IRX-1 | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC023667.2 | |
IRX3 | |
IRX3 (AAH23667.1, 1 a.a. ∼ 501 a.a) full-length human protein. | |
RUO | |
Yes | |
79191 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.