Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISGF3γ Mouse anti-Human, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610285

Catalog No. BDB610285


Add to cart

Description

Description

ISGF3γ is a nuclear DNA-binding protein. ISGF3 (Interferon Stimulated Gene Factor-3) is the primary transcription activator induced by the binding of interferon α (IFNα) to cell surface receptors. The functional ISGF3 is a complex of four polypeptides of 113kDa (Stat2), 91kDa (Stat1α), 84kDa (Stat1β), and 48kDa. The first three polypeptides (also known as ISGF3γ) each contain SH2 and SH3 domains. In response to either IFNα, IFNγ, EGF, PDGF, or CSF-1 binding their respective receptors, the cytoplasmic ISGF3α subunits are tyrosine-phosphorylated and translocated to the nucleus where they form an active complex with the 48kDa protein (ISGF3γ). This complex is responsible for modulating the transcription of interferon-stimulated genes (ISGs). Activated ISGF3α stabilizes ISGF3?-DNA interactions. This results in a ternary protein-DNA complex with a 25-fold greater stability than ISGF3γ-DNA. Thus, growth factors and cytokines activate a common signal transduction pathway that leads to the phosphorylation and nuclear translocation of a group of latent cytoplasmic transcription factors.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

ISGF3γ
6
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Interferon Stimulated Gene Factor-3
Affinity Purified
Human ISGF3γ (p48) aa.126-351
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit