Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISM1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524968

Catalog No. PIPA524968


Add to cart

Specifications

Specifications

ISM1
Polyclonal
PBS with 0.09% sodium azide
B1AKI9, A2ATD1
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1, isthmin-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140862, 319909
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ISM1
Liquid
ISM1
KLH conjugated synthetic peptide between 435-464 amino acids from the C-terminal region of human ISM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit