Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526061

Catalog No. PIPA526061


Add to cart

Description

Description

ITGA2 id a protein that belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane glycoproteins composed of a distinct alpha chain and a common beta chain. They are found on a wide variety of cell types including, T cells, fibroblasts and platelets. Integrins are involved in cell adhesion and also participate in cell-surface mediated signalling.
Specifications

Specifications

ITGA2
Polyclonal
Unconjugated
ITGA2
Liquid
ITGA2
KLH conjugated synthetic peptide between 1023-1049 amino acids from the C-terminal region of human ITGA2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P17301
BR, CD49B, GPIa, VLA-2, VLAA2, ITGA2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3673
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit