Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD103 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523996

Catalog No. PIPA523996


Add to cart

Description

Description

Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. ITGAE encodes an I-domain-containing alpha integrin that undergoes post-translational cleavage in the extracellular domain, yielding disulfide-linked heavy and light chains. In combination with the beta 7 integrin, this protein forms the E-cadherin binding integrin known as the human mucosal lymphocyte-1 antigen. This protein is preferentially expressed in human intestinal intraepithelial lymphocytes (IEL), and in addition to a role in adhesion, it may serve as an accessory molecule for IEL activation.
Specifications

Specifications

CD103
Polyclonal
PBS with 0.09% sodium azide
P38570
CD103, HUMINAE, HML-1 antigen, antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, integrin alpha-E, integrin alpha-IEL, mucosal lymphocyte 1 antigen
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3682
Western Blot
Unconjugated
ITGAE
Liquid
ITGAE
KLH conjugated synthetic peptide between 1146-1173 amino acids from the C-terminal region of human ITGAE
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit