Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127604

Catalog No. PIPA127604

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Integrin alpha-X/beta-2 is a receptor for fibrinogen. It recognizes the sequence G-P-R in fibrinogen. It mediates cell-cell interaction during inflammatory responses. It is especially important in monocyte adhesion and chemotaxis. ITGAX encodes the integr

PA1-27604 detects ITGAX from human samples.PA1-27604 may be successfully used in Western blot applications.The PA1-27462 immunogen is a synthetic peptide corresponding to human Integrin alpha-X.
Specifications

Specifications

Integrin alpha X/CD11c
Polyclonal
PBS with no preservative
P20702
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Chicken
IgY
50μg
Store at -20°C
Primary
3687
Western Blot
Unconjugated
ITGAX
Purified
ITGAX
Synthetic peptide corresponding to human Integrin alpha-X.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit