Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD104, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127608

Catalog No. PIPA127608

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Integrin alpha-6/beta-4 is a receptor for laminin. It plays a critical structural role in the hemidesmosome of epithelial cells.

PA1-27608 detects ITGB4 from human samples.PA1-27608 may be successfully used in Western blot applications.The PA1-27453 immunogen is a synthetic peptide corresponding to human Integrin beta-4.
Specifications

Specifications

Integrin beta-4/CD104
Polyclonal
PBS with no preservative
P16144
CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, ITGB4
Chicken
IgY
50μg
Store at -20°C
Primary
3691
Western Blot
Unconjugated
ITGB4
Purified
ITGB4
Synthetic peptide corresponding to human Integrin beta-4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit