Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITIH2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA513670
Description
This antibody is predicted to react with porcine based on sequence homology.
ITIH2 may act as a carrier of hyaluro in serum or as a binding protein between hyaluro and other matrix protein, including those on cell surfaces in tissues to regulate the localization, synthesis and degradation of hyaluro which are essential to cells undergoing biological processes.Specifications
ITIH2 | |
Polyclonal | |
ITIH2 | |
Inter-alpha-trypsin inhibitor heavy chain H2; Inter-alpha-inhibitor heavy chain 2; ITI heavy chain H2; Inter-alpha-trypsin inhibitor complex component II; Serum-derived hyaluro-associated protein; SHAP; IGHEP2; ITIH2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3698 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
P19823 | |
ITIH2 | |
KLH conjugated synthetic peptide between 858-886 amino acids from the C-terminal region of human ITIH2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title