Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ITIH4, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178286

Catalog No. NBP178286


Add to cart

Description

Description

ITIH4 Polyclonal antibody specifically detects ITIH4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

ITIH4
0.5 mg/mL
Western Blot 0.03 - 0.1 μg/mL, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:16000
Q14624
DKFZp686G21125, H4P, heavy chain-like, 1, IHRP, inter-alpha (globulin) inhibitor H4 (plasma Kallikrein-sensitive glycoprotein), inter-alpha-trypsin inhibitor family heavy chain-related protein, inter-alpha-trypsin inhibitor heavy chain H4, ITI-HC4, ITIHL1, PK-120, plasma kallikrein-sensitive glycoprotein 120, PRO1851
Goat
IgG
Immunogen affinity purified
RUO
Primary
3700
Human
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
ITIH4
Peptide with sequence C-SDDGRRTLRVQGND corresponding to C-Terminus according to NP_002209.2, NP_001159921.1.
103 kDa
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_002209.2; NP_001159921.1).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only