Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITIH5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524445

Catalog No. PIPA524445


Add to cart

Description

Description

This gene encodes a heavy chain component of one of the inter-alpha-trypsin inhibitor (ITI) family members. ITI proteins are involved in extracellular matrix stabilization and in the prevention of tumor metastasis. They are also structurally related plasma serine protease inhibitors and are composed of a light chain and varying numbers of heavy chains. This family member is thought to function as a tumor suppressor in breast and thyroid cancers. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

ITIH5
Polyclonal
PBS with 0.09% sodium azide
Q86UX2
ITI-HC5, ITI heavy chain H5, inter-alpha (globulin) inhibitor H5, inter-alpha-inhibitor heavy chain 5, inter-alpha-trypsin inhibitor heavy chain H5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80760
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ITIH5
Liquid
ITIH5
KLH conjugated synthetic peptide between 578-606 amino acids from the C-terminal region of human ITIH5
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit