Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITM2B, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human ITM2B protein.

Manufacturer:  Abnova Corporation H00009445D03P

Catalog No. 89-002-972


Add to cart

Description

Description

Sequence: MVKVTFNSALAQKETKKDEPKSGEEALIIPPDAVAVDCKDPDDVVPVGQRRAWCWCMCFGLAFMLAGVILGGAYLYKYFALQPDDVYYCGIKYIKDDVILNEPSADAPAALYQTIEENIKIFEEEEVEFISVPVPEFADSDPANIVHDFNKKLTAYLDLNLDKCYVIPLNTSIVMPPRNLLELLINIKAGTYLPQSYLIHEHMVITDRIENIDHLGFFIYRLCHDKETYKLQRRETIKGIQKREASNCFAIRHFENKFAVETLICS
Specifications

Specifications

ITM2B
Unconjugated
In 1X PBS, pH 7.4
BC016148.1
ABRI|BRI|BRI2|BRICD2B|E25B|E3-16|FBD
Rabbit
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine
Western Blot
Rabbit polyclonal antibody raised against a full-length human ITM2B protein.
ITM2B
Purified
ITM2B
ITM2B (AAH16148.1, 1 a.a. ~ 266 a.a) full-length human protein.
RUO
Primary
9445
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit