Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITM2B, Mouse, Clone: 1A10, Abnova™

Mouse monoclonal antibody raised against a partial recombinant ITM2B.

Manufacturer:  Abnova Corporation H00009445M05

Catalog No. 89-002-745


Add to cart

Description

Description

Sequence: VHDFNKKLTAYLDLNLDKCYVIPLNTSIVMPPRNLLELLINIKAGTYLPQSYLIHEHMVITDRIENIDHLGFFIYRLCHDKETYKLQRRETIKGIQKREA
Specifications

Specifications

ITM2B,
1A10
Mouse monoclonal antibody raised against a partial recombinant ITM2B.
ITM2B
Liquid
ITM2B
ITM2B (NP_068839, 146 a.a. ~ 245 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
In 1X PBS, pH 7.4
NM_021999
ABRI|BRI|BRI2|BRICD2B|E25B|E3-16|FBD
Mouse
IgG2a Kappa
RUO
Primary
9445
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit