Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITPA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524867

Catalog No. PIPA524867

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken, mouse, rat and Xenopus based on sequence homology.

ITPA hydrolyzes inosine triphosphate and deoxyinosine triphosphate to the monophosphate nucleotide and diphosphate. The encoded protein, which is a member of the HAM1 NTPase protein family, is found in the cytoplasm and acts as a homodimer. Defects in the encoded protein can result in inosine triphosphate pyrophosphorylase deficiency. Two transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

ITPA
Polyclonal
PBS with 0.09% sodium azide
Q9BY32
C20orf37, HLC14-06-P, ITPase, dJ794I6.3, My049 protein, NTPase, inosine triphosphatase-A, inosine triphosphate pyrophosphatase, inosine triphosphate pyrophosphohydrolase, non-canonical purine NTP pyrophosphatase, non-standard purine NTP pyrophosphatase, nucleoside triphosphate diphosphatase, nucleoside-
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3704
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
ITPA
Liquid
ITPA
KLH conjugated synthetic peptide between 24-51 amino acids from the N-terminal region of human ITPA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit