Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAB1/CSN5 Mouse, Unlabeled, Clone: 42, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611618

Catalog No. BDB611618


Add to cart

Description

Description

JAB1 (Jun activation domain-binding protein-1) was isolated in a yeast two- hybrid screen using the c-Jun N-terminal activation domain. Binding of JAB1 to Jun potentiates activator protein transcription factor (AP-1) target gene transcription. JAB1 interacts with c-Jun and JunD, but not with JunB or v-Jun. It also interacts with other transcriptional regulatory complexes, such as the activator of stromelysin 1 gene transcription complex (RNF4/SPBP) and the steroid receptor coactivator-1 complex. Additionally, JAB1 is the CSN5 subunit of the COP9 signalosome, which contains multiple proteins with homologies to proteins present in the 19S subunit of the proteosome. Interestingly, JAB1 has been implicated in the instigation of p27kip1 and rat lutropin/choriogonadotropin receptor degradation. JAB1 has also been found to be involved in integrin signaling cascades, because it colocalizes with the integrin LFA-1 receptor and is translocated to the nucleus, where it enhances transactivation of the AP-1- dependent promoter following integrin stimulation. Thus, JAB1 is a multifunctional protein that regulates both gene transcription and protein degradation.

Host Species: Mouse
Clone: 42
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human JAB1 aa. 234-334

Immunofluorescence, Western Blotting

Specifications

Specifications

JAB1/CSN5
42
Unconjugated
Affinity Purified
Human JAB1 aa. 234-334
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit