Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jagged 1 Mouse anti-Human, Mouse, Rat, Clone: 188331, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen Jagged 1
Immunogen NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504
Clone 188331
Applications Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1277SP
View Documents
R&D Systems
MAB1277SP
25 ug
Each for $114.50
Add to cart
 
MAB1277100
View Documents
R&D Systems
MAB1277100
100 ug
Each for $377.00
Add to cart
 
MAB1277500
View Documents
R&D Systems
MAB1277500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Jagged 1 Monoclonal antibody MAB1277 Specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications

Specifications

Jagged 1
188331
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2b
182
RUO
NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504
Flow Cytometry, CyTOF
Purified
P78504
Monoclonal
Primary
Detects human, mouse, and rat Jagged 1. In direct ELISAs, no cross-reactivity with recombinant human Jagged 2 is observed. Stains human Jagged 1 transfectants but not the parental cell line or human Jagged 2 transfectants in flow cytometry.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit