Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Jagged 2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178010

Catalog No. NBP178010


Add to cart

Description

Description

Jagged 2 Polyclonal antibody specifically detects Jagged 2 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Jagged 2
0.9 mg/ml
Western Blot 1:500-1:2000, ELISA 1:10000-1:50000
Q9QYE5
HJ2, jagged 2, Jagged2, protein jagged-2, SER2
Rabbit
IgG
0.1 mg
Cell Cycle and Replication, Stem Cell Signaling Pathway
Primary
3714
Mouse
ELISA, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
JAG2
Anti-Jagged 2 [RABBIT) Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 111-128 of Mouse Jagged 2. Jagged 2 is a Notch ligand, which shows distinct expression patterns in the developing and adult brain that overlaps that of Notch1. Expected to cross-react with rat Jagged 2.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Expected to cross-react with rat Jagged-2. Jagged 2 is a Notch ligand, which shows distinct expression patterns in the developing and adult brain that overlaps that of Notch1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only