Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-JAK1 (Tyr1022, Tyr1023), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA128444

Catalog No. PIPA128444

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Janus Kinase 1 (JAK1) is one of a number of related tyrosine kinases involved in cytoplasmic signal transduction. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at tyrosine

PA1-28444 detects JAK1 from human samples.PA1-28444 has been successfully used in Western blot applications.The PA1-28444 immunogen is: Synthetic phospho peptide (Human). The sequence is conserved in mouse and pig.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-JAK1 pTyr1022 + pTyr1023
Polyclonal
JAK1
Tyrosine-protein kinase JAK1, Janus kinase 1, JAK1A, JAK1B
Rabbit
Antigen affinity chromatography
RUO
3716
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
P23458, P52332
JAK1
Synthetic phospho peptide (Human). The sequence is conserved in mouse and pig.
50 μL
Primary
Human, Mouse
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit