Learn More
JAK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA527583
Catalog No. PIPA527583
Add to cart
Description
Description
PA5-27583 targets JAK1 in IF and WB applications and shows reactivity with Human samples. The PA5-27583 immunogen is recombit fragment corresponding to a region within amino acids 744 and 1067 of Human JAK1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Janus kinase 1 (JAK1), is a member of a new class of protein-tyrosine kinases (PTK) characterized by the presence of a second phosphotransferase-related domain immediately N-terminal to the PTK domain. The second phosphotransferase domain bears all the hallmarks of a protein kinase, although its structure differs significantly from that of the PTK and threonine/serine kinase family members. JAK1 is a large, widely expressed membrane-associated phosphoprotein. JAK1 is involved in the interferon-alpha/beta and -gamma signal transduction pathways. The reciprocal interdependence between JAK1 and TYK2 activities in the interferon-alpha pathway, and between JAK1 and JAK2 in the interferon-gamma pathway, may reflect a requirement for these kinases in the correct assembly of interferon receptor complexes. These kinases couple cytokine ligand binding to tyrosine phosphorylation of various known signaling proteins and of a unique family of transcription factors termed the signal transducers and activators of transcription, or STATs.
Specifications
Specifications
JAK1
Polyclonal
Unconjugated
JAK1
Liquid
JAK1
Recombit fragment corresponding to a region within amino acids 744 and 1067 of Human JAK1
Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
5.94 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P23458, P52332
JAK1A, JAK1B, JTK3, tyrosine-protein kinase JAK1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
16451, 3716