Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-JAM-B/VE-JAM, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP136746

Catalog No. NBP136746

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

JAM-B/VE-JAM
0.5 mg/mL
Western Blot 0.5 - 1 ug/ml, Peptide ELISA Detection limit 1:4000
NP_067042.1
C21orf43, CD322 antigen, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAMVEJAMCD322
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region.
Western Blot, ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
JAM2
Peptide with sequence C-QRKGYFSKETSFQ corresponding to internal region according to NP_067042.1.
Immunogen affinity purified
RUO
Primary
58494
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only