Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-JAMP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236446SS

 View more versions of this product

Catalog No. NBP236446SS


Add to cart

Description

Description

JAMP Polyclonal antibody specifically detects JAMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

JAMP
1.0 mg/mL
Western Blot 2 ug/ml, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 10 ug/ml
Q9P055
C14orf100, chromosome 14 open reading frame 100, HSPC213, HSPC327, JAMPCDA06, JNK1/MAPK8-associated membrane protein, JNK1-associated membrane protein, JNK-associated membrane protein, Jun N-terminal kinase 1-associated membrane protein, Medulloblastoma antigen MU-MB-50.4
Rabbit
IgG
0.025 ml
Unfolded Protein Response (UPR)
Primary
51528
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.05% Sodium Azide
Affinity Purified
JKAMP
A synthetic peptide made to an internal portion of the human JAMP protein (between amino acids 20-150) [UniProt Q9P055]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only